The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142883.1 | NP_001136355.1 | 4 - 5 | 954 | 131 |
NM_054111.4 | NP_473452.2 | 3 - 4 | 825 | 131 | |
XM_005248842.3 | XP_005248899.1 | 4 - 5 | 734 | 131 | |
XM_011514295.2 | XP_011512597.1 | 3 - 4 | 837 | 131 | |
GenBank mRNA | AF393812.1 | - | 2 - 3 | 520 | 131 |
AK056586.2 | - | 3 - 4 | 744 | 130 | |
AK223572.1 | - | 3 - 4 | 748 | 131 | |
AK289935.1 | - | 4 - 5 | 874 | 131 | |
BC089389.1 | - | 3 - 4 | 825 | 131 |