Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYT1L
Entrez Gene ID
23040
Gene Name
myelin transcription factor 1 like
Gene Aliases
MRD39, NZF1, ZC2H2C2, ZC2HC4B
Chromosome Location
Chr.2: 1789113 - 2331388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303052.1 NP_001289981.1 5 - 6 863 73
NM_015025.3 NP_055840.2 5 - 6 863 73
XM_011510318.1 XP_011508620.1 5 - 6 925 73
XM_011510319.1 XP_011508621.1 5 - 6 1035 73
XM_011510320.2 XP_011508622.1 5 - 6 966 73
XM_011510321.2 XP_011508623.1 5 - 6 916 73
XM_011510322.2 XP_011508624.1 5 - 6 729 73
XM_011510323.1 XP_011508625.1 4 - 5 648 73
XM_011510324.1 XP_011508626.1 5 - 6 925 73
XM_011510325.1 XP_011508627.1 5 - 6 925 73
XM_011510326.1 XP_011508628.1 5 - 6 925 73
XM_011510327.1 XP_011508629.1 5 - 6 925 73
XM_011510328.2 XP_011508630.1 5 - 6 925 73
XM_011510331.2 XP_011508633.1 5 - 6 925 73
XM_011510332.2 XP_011508634.1 5 - 6 925 73
XM_017003604.1 XP_016859093.1 6 - 7 1014 73
XM_017003605.1 XP_016859094.1 5 - 6 593 73
XM_017003606.1 XP_016859095.1 5 - 6 1035 73
XM_017003607.1 XP_016859096.1 5 - 6 966 73
XM_017003608.1 XP_016859097.1 5 - 6 925 73
XM_017003609.1 XP_016859098.1 5 - 6 910 73
XM_017003610.1 XP_016859099.1 6 - 7 1009 73
XM_017003611.1 XP_016859100.1 5 - 6 925 73
XM_017003612.1 XP_016859101.1 5 - 6 1035 73
XM_017003613.1 XP_016859102.1 5 - 6 1035 73
XM_017003614.1 XP_016859103.1 5 - 6 874 73
XM_017003615.1 XP_016859104.1 5 - 6 925 73
XM_017003616.1 XP_016859105.1 5 - 6 925 73
XM_017003617.1 XP_016859106.1 5 - 6 925 73
XM_017003618.1 XP_016859107.1 5 - 6 925 73
XM_017003619.1 XP_016859108.1 5 - 6 1035 73
XM_017003621.1 XP_016859110.1 5 - 6 925 73
XM_017003622.1 XP_016859111.1 5 - 6 925 73
GenBank mRNA AB029029.2 - 5 - 6 804 73
AF036943.1 - 5 - 6 758 73
AK299571.1 - 1 - 2 80 73
AK307797.1 - 5 - 6 861 73
BC109113.1 - 3 - 4 384 73
BC137272.1 - 5 - 6 666 73
BC137273.1 - 5 - 6 666 73
BC150281.1 - 5 - 6 804 73

View Full Product Details