Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYT1L
Entrez Gene ID
23040
Gene Name
myelin transcription factor 1 like
Gene Aliases
MRD39, NZF1, ZC2H2C2, ZC2HC4B
Chromosome Location
Chr.2: 1789113 - 2331388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303052.1 NP_001289981.1 8 - 9 1021 110
NM_015025.3 NP_055840.2 8 - 9 1021 110
XM_011510318.1 XP_011508620.1 8 - 9 1083 110
XM_011510319.1 XP_011508621.1 8 - 9 1193 110
XM_011510320.2 XP_011508622.1 8 - 9 1124 110
XM_011510321.2 XP_011508623.1 8 - 9 1074 110
XM_011510322.2 XP_011508624.1 8 - 9 887 110
XM_011510323.1 XP_011508625.1 7 - 8 806 110
XM_011510324.1 XP_011508626.1 8 - 9 1083 110
XM_011510325.1 XP_011508627.1 8 - 9 1083 110
XM_011510326.1 XP_011508628.1 8 - 9 1083 110
XM_011510327.1 XP_011508629.1 8 - 9 1083 110
XM_011510328.2 XP_011508630.1 8 - 9 1083 110
XM_011510331.2 XP_011508633.1 8 - 9 1083 110
XM_011510332.2 XP_011508634.1 8 - 9 1083 110
XM_017003604.1 XP_016859093.1 9 - 10 1172 110
XM_017003605.1 XP_016859094.1 8 - 9 751 110
XM_017003606.1 XP_016859095.1 8 - 9 1193 110
XM_017003607.1 XP_016859096.1 8 - 9 1124 110
XM_017003608.1 XP_016859097.1 8 - 9 1083 110
XM_017003609.1 XP_016859098.1 8 - 9 1068 110
XM_017003610.1 XP_016859099.1 9 - 10 1167 110
XM_017003611.1 XP_016859100.1 8 - 9 1083 110
XM_017003612.1 XP_016859101.1 8 - 9 1193 110
XM_017003613.1 XP_016859102.1 8 - 9 1193 110
XM_017003614.1 XP_016859103.1 8 - 9 1032 110
XM_017003615.1 XP_016859104.1 8 - 9 1083 110
XM_017003616.1 XP_016859105.1 8 - 9 1083 110
XM_017003617.1 XP_016859106.1 8 - 9 1083 110
XM_017003618.1 XP_016859107.1 8 - 9 1083 110
XM_017003619.1 XP_016859108.1 8 - 9 1193 110
XM_017003621.1 XP_016859110.1 8 - 9 1083 110
XM_017003622.1 XP_016859111.1 8 - 9 1083 110
GenBank mRNA AB029029.2 - 8 - 9 962 110
AF036943.1 - 8 - 9 916 110
BC137272.1 - 8 - 9 824 110
BC137273.1 - 8 - 9 824 110
BC150281.1 - 8 - 9 962 110

View Full Product Details