Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYT1L
Entrez Gene ID
23040
Gene Name
myelin transcription factor 1 like
Gene Aliases
MRD39, NZF1, ZC2H2C2, ZC2HC4B
Chromosome Location
Chr.2: 1789113 - 2331388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303052.1 NP_001289981.1 10 - 11 2351 86
XM_011510318.1 XP_011508620.1 10 - 11 2413 86
XM_011510319.1 XP_011508621.1 10 - 11 2523 86
XM_011510320.2 XP_011508622.1 10 - 11 2454 86
XM_011510321.2 XP_011508623.1 10 - 11 2404 86
XM_011510322.2 XP_011508624.1 10 - 11 2217 86
XM_011510323.1 XP_011508625.1 9 - 10 2136 86
XM_011510324.1 XP_011508626.1 10 - 11 2413 86
XM_011510326.1 XP_011508628.1 10 - 11 2413 86
XM_011510327.1 XP_011508629.1 10 - 11 2413 86
XM_011510328.2 XP_011508630.1 10 - 11 2413 86
XM_011510331.2 XP_011508633.1 10 - 11 2413 86
XM_011510332.2 XP_011508634.1 10 - 11 2413 86
XM_017003604.1 XP_016859093.1 11 - 12 2502 86
XM_017003605.1 XP_016859094.1 10 - 11 2081 86
XM_017003608.1 XP_016859097.1 10 - 11 2413 86
XM_017003609.1 XP_016859098.1 10 - 11 2398 86
XM_017003610.1 XP_016859099.1 11 - 12 2497 86
XM_017003615.1 XP_016859104.1 10 - 11 2413 86
XM_017003616.1 XP_016859105.1 10 - 11 2413 86
XM_017003620.1 XP_016859109.1 3 - 4 1403 86
XM_017003621.1 XP_016859110.1 10 - 11 2413 86
XM_017003622.1 XP_016859111.1 10 - 11 2413 86
GenBank mRNA AF036943.1 - 10 - 11 2246 86
AK299571.1 - 6 - 7 1568 86
AL133024.1 - 1 - 2 446 86

View Full Product Details