The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294339.1 | NP_001281268.1 | 3 - 4 | 726 | 61 |
XM_017000242.1 | XP_016855731.1 | 3 - 4 | 701 | 61 | |
GenBank mRNA | AK303696.1 | - | 4 - 5 | 505 | 61 |
L29216.1 | - | 3 - 4 | 526 | 61 |