The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166284.1 | NP_001159756.1 | 3 - 4 | 328 | 64 |
NM_001166285.1 | NP_001159757.1 | 6 - 7 | 685 | 64 | |
NM_006429.3 | NP_006420.1 | 5 - 6 | 589 | 64 | |
NR_029403.1 | 4 - 5 | 440 | 64 | ||
XM_011532478.2 | XP_011530780.1 | 6 - 7 | 619 | 64 | |
XM_011532479.1 | XP_011530781.1 | 5 - 6 | 522 | 64 | |
GenBank mRNA | AF026292.1 | - | 5 - 6 | 514 | 64 |
AK098549.1 | - | 5 - 6 | 507 | 64 | |
AK222478.1 | - | 5 - 6 | 497 | 64 | |
AK297846.1 | - | 6 - 7 | 613 | 64 | |
AK298153.1 | - | 3 - 4 | 256 | 64 | |
AK302078.1 | - | 4 - 5 | 368 | 64 | |
BC019296.1 | - | 5 - 6 | 502 | 64 | |
BC088351.1 | - | 5 - 6 | 544 | 64 | |
BM918058.1 | - | - | 565 | 64 | |
CR456719.1 | - | 4 - 5 | 446 | 64 | |
CR536511.1 | - | 4 - 5 | 446 | 64 | |
U83843.1 | - | 4 - 5 | 410 | 64 |