Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC5L
Entrez Gene ID
988
Gene Name
cell division cycle 5 like
Gene Aliases
CDC5, CDC5-LIKE, CEF1, PCDC5RP, dJ319D22.1
Chromosome Location
Chr.6: 44387514 - 44450424 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253.3 NP_001244.1 3 - 4 616 124
GenBank mRNA AB007892.2 - 3 - 4 388 124
AK128737.1 - 3 - 4 424 124
BC001568.1 - 3 - 4 405 124
BG497404.1 - - 568 124
D85423.1 - 2 - 3 285 124
U86753.1 - 3 - 4 306 124

View Full Product Details