The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007359.4 | NP_031385.2 | 5 - 6 | 907 | 77 |
XM_005257163.1 | XP_005257220.1 | 5 - 6 | 685 | 77 | |
GenBank mRNA | AK292425.1 | - | 5 - 6 | 846 | 77 |
AK296682.1 | - | 5 - 6 | 848 | 77 | |
BC030008.1 | - | 5 - 6 | 642 | 77 | |
BC044656.1 | - | 5 - 6 | 790 | 77 | |
BC050526.1 | - | 5 - 6 | 810 | 77 | |
X80199.2 | - | 5 - 6 | 845 | 77 |