Product Details

Assay Reference Genome
Location

Chr.7:100406701 on build GRCh38
Cytoband
7q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
55063
Gene Symbol
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Location
Chr.7:100400826-100429239 on build GRCh38
Assay Gene Location
Overlaps Intron 13 - Exon 13
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ZCWPW1 NM_001258008.1 NP_001244937.1
NM_017984.4 NP_060454.3
XM_005250479.2 XP_005250536.1
XM_005250480.2 XP_005250537.1
XM_006716035.3 XP_006716098.1
XM_006716036.2 XP_006716099.1
XM_006716037.3 XP_006716100.1
XM_006716038.3 XP_006716101.1
XM_006716040.3 XP_006716103.1
XM_017012379.1 XP_016867868.1
XM_017012380.1 XP_016867869.1
AK000919.1 BAA91424.1
AK093038.1 BAC04028.1
AK300745.1
AK302108.1
AL136735.1 CAB66669.1
BC002725.1 AAH02725.1
KU178677.1
KU178678.1
KU178679.1
KU178680.1
KU178681.1
KU178682.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2734888 Chr7:99975623 - 101290285 on Build GRCh38 Deletion LOC102724094 TRIP6 COPS6 MOSPD3 PLOD3 ZKSCAN1 AZGP1P1 MBLAC1 SERPINE1 AGFG2 ZNHIT1 ZASP MIR4653 GAL3ST4 VGF GATS AP4M1 PMS2P1 EPO GNB2 EPHB4 MIR6840 MIR4658 PCOLCE ZSCAN21 CNPY4 FIS1 PVRIG2P UFSP1 TAF6 STAG3L5P ZCWPW1 CLDN15 POP7 SLC12A9 C7orf43 LRCH4 LOC105375429 PCOLCE-AS1 MIR25 ZAN MUC3A STAG3L5P-PVRIG2P-PILRB MCM7 AP1S1 ACHE SAP25 ACTL6B MUC17 MOGAT3 PILRB TSC22D4 PILRA TFR2 MEPCE SRRT C7orf61 PVRIG MIR106B MIR6875 NAT16 GPC2 LAMTOR4 FBXO24 MUC12 NYAP1 GIGYF1 PPP1R35 TRIM56 SPDYE3 ZNF3 STAG3 MIR93 AZGP1
nsv831071 Chr7:100320402 - 100515790 on Build GRCh38 Loss NYAP1 STAG3L5P-PVRIG2P-PILRB MIR6840 PPP1R35 PILRB PVRIG2P STAG3L5P TSC22D4 ZCWPW1 PILRA MEPCE SPDYE3 C7orf61 PMS2P1

View Full Product Details