The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040092.2 | NP_001035181.1 | 6 - 7 | 663 | 78 |
NM_001130863.2 | NP_001124335.1 | 6 - 7 | 663 | 78 | |
NM_006209.4 | NP_006200.3 | 6 - 7 | 663 | 78 | |
XM_006716584.1 | XP_006716647.1 | 6 - 7 | 641 | 78 | |
XM_006716585.1 | XP_006716648.1 | 6 - 7 | 641 | 78 | |
XM_006716587.1 | XP_006716650.1 | 6 - 7 | 641 | 78 | |
XM_017013571.1 | XP_016869060.1 | 6 - 7 | 697 | 78 | |
XM_017013572.1 | XP_016869061.1 | 6 - 7 | 641 | 78 | |
XM_017013573.1 | XP_016869062.1 | 6 - 7 | 697 | 78 | |
XM_017013574.1 | XP_016869063.1 | 6 - 7 | 697 | 78 | |
XM_017013575.1 | XP_016869064.1 | 6 - 7 | 641 | 78 | |
GenBank mRNA | AK315253.1 | - | 6 - 7 | 636 | 78 |
BC034961.2 | - | 6 - 7 | 636 | 78 | |
D45421.1 | - | 6 - 7 | 610 | 78 | |
EU131011.2 | - | 6 - 7 | 577 | 78 | |
L35594.1 | - | 6 - 7 | 626 | 78 |