The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018711.4 | NP_061181.1 | 13 - 14 | 1638 | 103 |
GenBank mRNA | AK309780.1 | - | 4 - 5 | 383 | 103 |
AL359592.1 | - | 1 - 2 | 56 | 103 | |
BC033587.1 | - | 13 - 14 | 1438 | 103 | |
BC094722.1 | - | 13 - 14 | 1285 | 103 |