Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P2RX4
Entrez Gene ID
5025
Gene Name
purinergic receptor P2X 4
Gene Aliases
P2X4, P2X4R
Chromosome Location
Chr.12: 121209861 - 121234106 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK293599.1 - 3 - 4 394 125

View Full Product Details