Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASNS
Entrez Gene ID
440
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Gene Aliases
ASNSD, TS11
Chromosome Location
Chr.7: 97852117 - 97872542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178075.1 NP_001171546.1 7 - 8 1081 103
NM_001178076.1 NP_001171547.1 5 - 6 847 103
NM_001178077.1 NP_001171548.1 5 - 6 780 103
NM_001673.4 NP_001664.3 7 - 8 1155 103
NM_133436.3 NP_597680.2 7 - 8 1410 103
NM_183356.3 NP_899199.2 8 - 9 1433 103
GenBank mRNA AK000379.1 - 7 - 8 1378 103
AK302189.1 - 7 - 8 975 103
AK302242.1 - 5 - 6 728 103
AK316224.1 - 5 - 6 782 103
BC008723.2 - 8 - 9 1260 103
BC014621.2 - 7 - 8 1031 103
BT007113.1 - 5 - 6 904 103
M15798.1 - 6 - 7 951 103
M27396.1 - 7 - 8 1082 103

View Full Product Details