Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H3
Entrez Gene ID
23144
Gene Name
zinc finger CCCH-type containing 3
Gene Aliases
ZC3HDC3
Chromosome Location
Chr.8: 143437655 - 143549729 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015117.2 NP_055932.2 1 - 2 70 82
XM_006716536.3 XP_006716599.2 3 - 4 459 82
XM_011516943.2 XP_011515245.1 1 - 2 185 82
XM_011516944.2 XP_011515246.1 1 - 2 185 82
XM_017013248.1 XP_016868737.1 2 - 3 1090 82
XM_017013249.1 XP_016868738.1 2 - 3 632 82
GenBank mRNA BC038670.1 - 1 - 2 53 82
BM148116.1 - 1 - 2 73 82

View Full Product Details