The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005141.4 | NP_005132.2 | 2 - 3 | 349 | 73 |
GenBank mRNA | AK303788.1 | - | 3 - 4 | 267 | 73 |
AK309592.1 | - | 2 - 3 | 310 | 73 | |
AK312972.1 | - | 2 - 3 | 349 | 73 | |
BC007030.1 | - | 2 - 3 | 314 | 73 | |
BC070245.1 | - | 2 - 3 | 310 | 73 | |
BC106760.2 | - | 2 - 3 | 318 | 73 | |
BC107766.1 | - | 2 - 3 | 318 | 73 | |
EU668333.1 | - | 2 - 3 | 335 | 73 | |
J00129.1 | - | 2 - 3 | 286 | 73 |