The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184741.1 | NP_001171670.1 | 6 - 7 | 821 | 88 |
NM_005141.4 | NP_005132.2 | 6 - 7 | 998 | 88 | |
GenBank mRNA | AK303788.1 | - | 7 - 8 | 916 | 88 |
AK309592.1 | - | 6 - 7 | 943 | 88 | |
AK312972.1 | - | 6 - 7 | 998 | 88 | |
BC036007.1 | - | 6 - 7 | 789 | 88 | |
BC106760.2 | - | 6 - 7 | 967 | 88 | |
BC107766.1 | - | 6 - 7 | 967 | 88 | |
EU668333.1 | - | 6 - 7 | 984 | 88 | |
J00129.1 | - | 6 - 7 | 935 | 88 | |
J00133.1 | - | 2 - 3 | 179 | 88 |