The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145685.2 | NP_663718.1 | 1 - 2 | 821 | 58 |
GenBank mRNA | AJ297406.1 | - | 1 - 2 | 1375 | 58 |
AJ297407.1 | - | 1 - 2 | 829 | 58 | |
BC103861.2 | - | 1 - 2 | 129 | 58 | |
BC133655.1 | - | 1 - 2 | 129 | 58 |