The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324307.1 | NP_001311236.1 | 3 - 4 | 487 | 79 |
NM_001324308.1 | NP_001311237.1 | 2 - 3 | 412 | 79 | |
NM_002511.3 | NP_002502.2 | 2 - 3 | 913 | 79 | |
XM_011535846.1 | XP_011534148.1 | 4 - 5 | 543 | 79 | |
XM_017010901.1 | XP_016866390.1 | 2 - 3 | 512 | 79 | |
XM_017010902.1 | XP_016866391.1 | 2 - 3 | 449 | 79 | |
GenBank mRNA | BC095542.1 | - | 2 - 3 | 773 | 79 |
GU727635.1 | - | 2 - 3 | 772 | 79 | |
M73482.1 | - | 2 - 3 | 911 | 79 |