The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015423.2 | NP_056238.2 | 4 - 5 | 836 | 142 |
GenBank mRNA | AF136978.1 | - | 4 - 5 | 858 | 142 |
AF151057.1 | - | 4 - 5 | 856 | 142 | |
AF151838.1 | - | 4 - 5 | 856 | 142 | |
AF201943.1 | - | 4 - 5 | 805 | 142 | |
AF302110.1 | - | 4 - 5 | 856 | 142 | |
AK293362.1 | - | 3 - 4 | 597 | 142 | |
AK312529.1 | - | 4 - 5 | 740 | 142 | |
AL050073.1 | - | 3 - 4 | 397 | 142 | |
BC015470.1 | - | 4 - 5 | 727 | 142 | |
BC016728.1 | - | 4 - 5 | 756 | 142 |