The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271004.1 | NP_001257933.1 | 3 - 4 | 780 | 60 |
GenBank mRNA | AK075425.1 | - | 3 - 4 | 541 | 60 |
DA833200.1 | - | 3 - 4 | 541 | 60 |