Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OS9
Entrez Gene ID
10956
Gene Name
OS9, endoplasmic reticulum lectin
Gene Aliases
ERLEC2, OS-9
Chromosome Location
Chr.12: 57693955 - 57721557 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017956.2 NP_001017956.1 3 - 4 607 65
NM_001017957.2 NP_001017957.1 3 - 4 607 65
NM_001017958.2 NP_001017958.1 3 - 4 607 65
NM_001261420.1 NP_001248349.1 3 - 4 607 65
NM_001261421.1 NP_001248350.1 3 - 4 607 65
NM_001261423.1 NP_001248352.1 2 - 3 430 65
NM_006812.3 NP_006803.1 3 - 4 607 65
XM_005268581.1 XP_005268638.1 3 - 4 539 65
XM_006719200.1 XP_006719263.1 3 - 4 539 65
XM_006719201.1 XP_006719264.1 3 - 4 539 65
GenBank mRNA AB002806.1 - 3 - 4 441 65
AK129781.1 - 3 - 4 419 65
AK130733.1 - 3 - 4 417 65
AK291374.1 - 3 - 4 489 65
AK293435.1 - 2 - 3 242 65
AK296770.1 - 3 - 4 417 65
AK298532.1 - 3 - 4 417 65
AK303291.1 - 3 - 4 575 65
AL137691.1 - 1 - 2 345 65
AU132426.1 - 3 - 4 459 65
BC000532.2 - 3 - 4 412 65
BC007254.1 - 3 - 4 412 65
BC023513.2 - 3 - 4 402 65
CR456791.1 - 3 - 4 400 65
U41635.1 - 3 - 4 485 65

View Full Product Details