The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017956.2 | NP_001017956.1 | 4 - 5 | 687 | 62 |
NM_001017957.2 | NP_001017957.1 | 4 - 5 | 687 | 62 | |
NM_001017958.2 | NP_001017958.1 | 4 - 5 | 687 | 62 | |
NM_001261420.1 | NP_001248349.1 | 4 - 5 | 687 | 62 | |
NM_001261423.1 | NP_001248352.1 | 3 - 4 | 510 | 62 | |
NM_006812.3 | NP_006803.1 | 4 - 5 | 687 | 62 | |
XM_005268581.1 | XP_005268638.1 | 4 - 5 | 619 | 62 | |
XM_006719200.1 | XP_006719263.1 | 4 - 5 | 619 | 62 | |
XM_006719201.1 | XP_006719264.1 | 4 - 5 | 619 | 62 | |
GenBank mRNA | AB002806.1 | - | 4 - 5 | 521 | 62 |
AK129781.1 | - | 4 - 5 | 499 | 62 | |
AK130733.1 | - | 4 - 5 | 497 | 62 | |
AK291374.1 | - | 4 - 5 | 569 | 62 | |
AK293435.1 | - | 3 - 4 | 322 | 62 | |
AK296770.1 | - | 4 - 5 | 497 | 62 | |
AK303291.1 | - | 4 - 5 | 655 | 62 | |
AU132426.1 | - | 4 - 5 | 539 | 62 | |
BC000532.2 | - | 4 - 5 | 492 | 62 | |
BC007254.1 | - | 4 - 5 | 492 | 62 | |
BC023513.2 | - | 4 - 5 | 482 | 62 | |
CR456791.1 | - | 4 - 5 | 480 | 62 | |
U41635.1 | - | 4 - 5 | 565 | 62 |