The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172684.1 | NP_001166155.1 | 4 - 5 | 593 | 87 |
NM_001172685.1 | NP_001166156.1 | 3 - 4 | 351 | 87 | |
NM_052840.4 | NP_443072.3 | 4 - 5 | 831 | 87 | |
GenBank mRNA | AF401233.1 | - | 4 - 5 | 776 | 87 |
AF425606.1 | - | 4 - 5 | 592 | 87 | |
AK124858.1 | - | 3 - 4 | 385 | 87 | |
AK131098.1 | - | 6 - 7 | 1980 | 87 | |
AK294382.1 | - | 3 - 4 | 351 | 87 | |
AK296004.1 | - | 4 - 5 | 529 | 87 | |
BC030835.1 | - | 4 - 5 | 534 | 87 | |
BC033838.2 | - | 4 - 5 | 435 | 87 |