The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286434.1 | NP_001273363.1 | 6 - 7 | 698 | 87 |
NM_153356.2 | NP_699187.1 | 7 - 8 | 806 | 87 | |
XM_006720409.3 | XP_006720472.1 | 7 - 8 | 836 | 87 | |
XM_006720410.3 | XP_006720473.1 | 6 - 7 | 728 | 87 | |
XM_006720411.3 | XP_006720474.1 | 5 - 6 | 624 | 87 | |
XM_011521281.2 | XP_011519583.1 | 7 - 8 | 835 | 87 | |
XM_011521282.2 | XP_011519584.1 | 7 - 8 | 835 | 87 | |
XM_011521283.2 | XP_011519585.1 | 7 - 8 | 835 | 87 | |
XM_011521284.2 | XP_011519586.1 | 6 - 7 | 615 | 87 | |
GenBank mRNA | AB449907.1 | - | 6 - 7 | 574 | 87 |
BC033516.1 | - | 7 - 8 | 765 | 87 |