The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000048.3 | NP_000039.2 | 1 - 2 | 73 | 65 |
GenBank mRNA | BX328836.2 | - | 1 - 2 | 93 | 65 |