The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006998.3 | NP_008929.2 | 1 - 2 | 283 | 107 |
GenBank mRNA | AK289477.1 | - | 1 - 2 | 338 | 107 |
BC000336.2 | - | 1 - 2 | 250 | 107 | |
BC003036.1 | - | 1 - 2 | 250 | 107 | |
BM511122.1 | - | 1 - 2 | 289 | 107 | |
Y16752.1 | - | 1 - 2 | 236 | 107 |