The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005914.3 | NP_005905.2 | 5 - 6 | 1545 | 126 |
NM_182746.2 | NP_877423.1 | 6 - 7 | 1429 | 126 | |
GenBank mRNA | AK022899.1 | - | 6 - 7 | 658 | 126 |
AK296913.1 | - | 5 - 6 | 637 | 126 | |
AK309812.1 | - | 6 - 7 | 499 | 126 | |
BC031061.1 | - | 5 - 6 | 702 | 126 | |
X74794.1 | - | 5 - 6 | 785 | 126 |