The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130848.1 | NP_001124320.1 | 12 - 13 | 2005 | 92 |
NM_004910.2 | NP_004901.2 | 12 - 13 | 2005 | 92 | |
XM_011545396.2 | XP_011543698.1 | 12 - 13 | 2021 | 92 | |
XM_017018586.1 | XP_016874075.1 | 12 - 13 | 2065 | 92 | |
GenBank mRNA | AF334584.1 | - | 12 - 13 | 1952 | 92 |
AK126944.1 | - | 2 - 3 | 610 | 92 | |
AK312886.1 | - | 11 - 12 | 1783 | 92 | |
AY429102.1 | - | 11 - 12 | 1779 | 92 | |
BC022230.1 | - | 12 - 13 | 1998 | 92 | |
X98654.1 | - | 11 - 12 | 1968 | 92 |