The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130848.1 | NP_001124320.1 | 3 - 4 | 522 | 92 |
NM_004910.2 | NP_004901.2 | 3 - 4 | 522 | 92 | |
XM_011545396.2 | XP_011543698.1 | 3 - 4 | 541 | 92 | |
XM_017018586.1 | XP_016874075.1 | 3 - 4 | 582 | 92 | |
GenBank mRNA | AF334584.1 | - | 3 - 4 | 469 | 92 |
AK312886.1 | - | 2 - 3 | 300 | 92 | |
AY429102.1 | - | 2 - 3 | 296 | 92 | |
BC022230.1 | - | 3 - 4 | 515 | 92 | |
X98654.1 | - | 2 - 3 | 485 | 92 |