Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSUH2
Entrez Gene ID
51066
Gene Name
ssu-2 homolog (C. elegans)
Gene Aliases
C3orf32, SSU-2, fls485
Chromosome Location
Chr.3: 8619400 - 8745035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256748.1 NP_001243677.1 10 - 11 912 74
NM_001256749.1 NP_001243678.1 10 - 11 933 74
NM_015931.2 NP_057015.1 10 - 11 1079 74
NR_046358.1 11 - 12 1160 74
XM_011533774.1 XP_011532076.1 18 - 19 2628 74
XM_017006510.1 XP_016861999.1 11 - 12 5393 74
XM_017006511.1 XP_016862000.1 17 - 18 2316 74
XM_017006512.1 XP_016862001.1 17 - 18 2485 74
XM_017006513.1 XP_016862002.1 16 - 17 2321 74
XM_017006514.1 XP_016862003.1 16 - 17 2227 74
XM_017006515.1 XP_016862004.1 18 - 19 3382 74
XM_017006516.1 XP_016862005.1 13 - 14 1743 74
XM_017006517.1 XP_016862006.1 13 - 14 1696 74
XM_017006518.1 XP_016862007.1 18 - 19 2607 74
XM_017006519.1 XP_016862008.1 11 - 12 5338 74
XM_017006520.1 XP_016862009.1 11 - 12 1689 74
XM_017006521.1 XP_016862010.1 14 - 15 1847 74
XM_017006522.1 XP_016862011.1 14 - 15 1738 74
XM_017006523.1 XP_016862012.1 12 - 13 1441 74
XM_017006524.1 XP_016862013.1 13 - 14 1679 74
XM_017006525.1 XP_016862014.1 16 - 17 2248 74
XM_017006526.1 XP_016862015.1 11 - 12 1327 74
XM_017006527.1 XP_016862016.1 17 - 18 2370 74
XM_017006528.1 XP_016862017.1 11 - 12 1366 74
XM_017006529.1 XP_016862018.1 11 - 12 1396 74
XM_017006530.1 XP_016862019.1 13 - 14 1829 74
XM_017006531.1 XP_016862020.1 12 - 13 1751 74
XM_017006533.1 XP_016862022.1 17 - 18 2403 74
XM_017006534.1 XP_016862023.1 17 - 18 2349 74
XM_017006538.1 XP_016862027.1 15 - 16 1946 74
XM_017006539.1 XP_016862028.1 10 - 11 1058 74
XM_017006540.1 XP_016862029.1 11 - 12 1237 74
XM_017006541.1 XP_016862030.1 8 - 9 879 74
XM_017006542.1 XP_016862031.1 8 - 9 857 74
GenBank mRNA AB024705.1 - 10 - 11 1052 74
AK057972.1 - 11 - 12 1156 74
AK093066.1 - 16 - 17 2109 74
AK122938.1 - 15 - 16 2677 74
AK225925.1 - 11 - 12 1156 74
AK300175.1 - 10 - 11 912 74
BC052614.1 - 10 - 11 916 74

View Full Product Details