The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018385.2 | NP_060855.2 | 5 - 6 | 840 | 67 |
GenBank mRNA | AK002163.1 | - | 5 - 6 | 573 | 67 |
AK225094.1 | - | 5 - 6 | 541 | 67 | |
AL136897.1 | - | 5 - 6 | 545 | 67 | |
BC015042.1 | - | 5 - 6 | 545 | 67 | |
BC040119.1 | - | 5 - 6 | 525 | 67 | |
BC058891.1 | - | 5 - 6 | 531 | 67 | |
BC068500.1 | - | 5 - 6 | 554 | 67 |