The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018385.2 | NP_060855.2 | 9 - 10 | 1587 | 55 |
XM_017006796.1 | XP_016862285.1 | 5 - 6 | 915 | 55 | |
GenBank mRNA | AK002163.1 | - | 9 - 10 | 1320 | 55 |
AK225094.1 | - | 9 - 10 | 1288 | 55 | |
AL136897.1 | - | 9 - 10 | 1292 | 55 | |
BC015042.1 | - | 9 - 10 | 1292 | 55 | |
BC040119.1 | - | 9 - 10 | 1272 | 55 | |
BC058891.1 | - | 9 - 10 | 1278 | 55 | |
BC068500.1 | - | 9 - 10 | 1301 | 55 |