The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016199.2 | NP_057283.1 | 1 - 2 | 55 | 80 |
GenBank mRNA | BI553911.1 | - | 1 - 2 | 60 | 80 |