The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006830.3 | NP_006821.1 | 2 - 3 | 276 | 102 |
GenBank mRNA | BC000462.2 | - | 2 - 3 | 224 | 102 |
BC003594.1 | - | 2 - 3 | 224 | 102 | |
D55636.1 | - | 2 - 3 | 238 | 102 | |
DW465790.1 | - | 2 - 3 | 281 | 102 |