Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRTAC1
Entrez Gene ID
55118
Gene Name
cartilage acidic protein 1
Gene Aliases
ASPIC, ASPIC1, CEP-68
Chromosome Location
Chr.10: 97854711 - 98030828 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206528.2 NP_001193457.1 1 - 2 382 147
NM_018058.6 NP_060528.3 1 - 2 382 147
XM_005269938.4 XP_005269995.1 1 - 2 175 147
XM_011539917.1 XP_011538219.1 1 - 2 175 147
XM_017016366.1 XP_016871855.1 1 - 2 175 147
XM_017016367.1 XP_016871856.1 1 - 2 175 147
GenBank mRNA AJ276171.1 - 1 - 2 175 147
AK057190.1 - 1 - 2 382 147
AK292084.1 - 1 - 2 144 147

View Full Product Details