Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPV5
Entrez Gene ID
56302
Gene Name
transient receptor potential cation channel subfamily V member 5
Gene Aliases
CAT2, ECAC1, OTRPC3
Chromosome Location
Chr.7: 142908101 - 142933723 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019841.6 NP_062815.3 8 - 9 1384 67
GenBank mRNA AF304464.1 - 8 - 9 1469 67
AJ271207.2 - 8 - 9 1120 67
AJ487965.2 - 8 - 9 1122 67

View Full Product Details