The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033050.5 | NP_149039.2 | 1 - 2 | 66 | 78 |
GenBank mRNA | AK290420.1 | - | 1 - 2 | 66 | 78 |
BC030948.1 | - | 1 - 2 | 60 | 78 |