The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178496.3 | NP_848591.2 | 1 - 2 | 531 | 72 |
GenBank mRNA | AK290447.1 | - | 1 - 2 | 226 | 72 |
BC013123.1 | - | 1 - 2 | 209 | 72 | |
BC036194.1 | - | 1 - 2 | 515 | 72 | |
BC045582.1 | - | 1 - 2 | 531 | 72 | |
BX648517.1 | - | 1 - 2 | 230 | 72 |