Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STX5
Entrez Gene ID
6811
Gene Name
syntaxin 5
Gene Aliases
SED5, STX5A
Chromosome Location
Chr.11: 62806860 - 62832091 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244666.1 NP_001231595.1 8 - 9 831 88
NM_003164.4 NP_003155.2 8 - 9 831 88
XM_011545223.2 XP_011543525.1 8 - 9 656 88
XM_011545224.2 XP_011543526.1 7 - 8 547 88
GenBank mRNA AK296675.1 - 7 - 8 547 88
AK313497.1 - 8 - 9 791 88
BC002645.1 - 8 - 9 781 88
BC012137.1 - 8 - 9 808 88
BT019646.1 - 7 - 8 512 88
BT019647.1 - 7 - 8 512 88
BX537426.1 - 8 - 9 829 88
U26648.1 - 7 - 8 538 88

View Full Product Details