Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLCO1C1
Entrez Gene ID
53919
Gene Name
solute carrier organic anion transporter family member 1C1
Gene Aliases
OATP-F, OATP1, OATP14, OATP1C1, OATPF, OATPRP5, SLC21A14
Chromosome Location
Chr.12: 20690505 - 20753386 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145945.1 NP_001139417.1 14 - 15 2116 95
NM_017435.4 NP_059131.1 14 - 15 2276 95
XM_005253394.2 XP_005253451.1 15 - 16 2052 95
XM_011520704.2 XP_011519006.1 15 - 16 2049 95
XM_017019486.1 XP_016874975.1 14 - 15 6971 95
XM_017019487.1 XP_016874976.1 13 - 14 6817 95
GenBank mRNA AF205076.1 - 14 - 15 2277 95
AF260704.1 - 14 - 15 1972 95
AK296236.1 - 14 - 15 2009 95
BC022461.2 - 14 - 15 2005 95

View Full Product Details