The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017643.2 | NP_060113.2 | 7 - 8 | 943 | 92 |
XM_005257467.4 | XP_005257524.1 | 6 - 7 | 888 | 92 | |
XM_005257468.4 | XP_005257525.1 | 6 - 7 | 881 | 92 | |
XM_011524922.2 | XP_011523224.1 | 6 - 7 | 890 | 92 | |
XM_011524923.2 | XP_011523225.1 | 6 - 7 | 887 | 92 | |
XM_011524924.2 | XP_011523226.1 | 6 - 7 | 883 | 92 | |
XM_011524925.1 | XP_011523227.1 | 7 - 8 | 1004 | 92 | |
XM_011524926.2 | XP_011523228.1 | 7 - 8 | 1108 | 92 | |
XM_011524928.2 | XP_011523230.1 | 6 - 7 | 1045 | 92 | |
XM_011524929.2 | XP_011523231.1 | 7 - 8 | 959 | 92 | |
XM_011524930.1 | XP_011523232.1 | 6 - 7 | 681 | 92 | |
XM_011524931.2 | XP_011523233.1 | 5 - 6 | 515 | 92 | |
XM_011524933.2 | XP_011523235.1 | 8 - 9 | 1065 | 92 | |
XM_011524934.2 | XP_011523236.1 | 6 - 7 | 889 | 92 | |
XM_011524935.2 | XP_011523237.1 | 6 - 7 | 882 | 92 | |
XM_011524936.1 | XP_011523238.1 | 6 - 7 | 681 | 92 | |
XM_017024782.1 | XP_016880271.1 | 7 - 8 | 1255 | 92 | |
GenBank mRNA | AK000062.1 | - | 3 - 4 | 275 | 92 |
AK124061.1 | - | 6 - 7 | 681 | 92 | |
BC034364.1 | - | 7 - 8 | 914 | 92 | |
BC101736.1 | - | 2 - 3 | 204 | 92 |