Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBTD1
Entrez Gene ID
54799
Gene Name
mbt domain containing 1
Gene Aliases
SA49P01
Chromosome Location
Chr.17: 51177419 - 51260511 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017643.2 NP_060113.2 10 - 11 1401 80
XM_005257467.4 XP_005257524.1 9 - 10 1346 80
XM_005257468.4 XP_005257525.1 9 - 10 1339 80
XM_011524922.2 XP_011523224.1 9 - 10 1348 80
XM_011524923.2 XP_011523225.1 9 - 10 1345 80
XM_011524924.2 XP_011523226.1 9 - 10 1341 80
XM_011524925.1 XP_011523227.1 10 - 11 1462 80
XM_011524926.2 XP_011523228.1 10 - 11 1566 80
XM_011524928.2 XP_011523230.1 9 - 10 1503 80
XM_011524929.2 XP_011523231.1 10 - 11 1417 80
XM_011524930.1 XP_011523232.1 9 - 10 1139 80
XM_011524931.2 XP_011523233.1 8 - 9 973 80
XM_011524933.2 XP_011523235.1 11 - 12 1523 80
XM_011524934.2 XP_011523236.1 9 - 10 1347 80
XM_011524935.2 XP_011523237.1 9 - 10 1340 80
XM_011524936.1 XP_011523238.1 9 - 10 1139 80
XM_017024782.1 XP_016880271.1 10 - 11 1713 80
GenBank mRNA AK000062.1 - 6 - 7 733 80
AK124061.1 - 9 - 10 1139 80
BC034364.1 - 10 - 11 1372 80
BC101736.1 - 5 - 6 662 80

View Full Product Details