The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012088.2 | NP_036220.1 | 2 - 3 | 448 | 64 |
XM_011527897.2 | XP_011526199.1 | 2 - 3 | 1048 | 64 | |
GenBank mRNA | AF091091.1 | - | 1 - 2 | 548 | 64 |
AJ243972.1 | - | 2 - 3 | 415 | 64 | |
BC014006.2 | - | 2 - 3 | 422 | 64 | |
BG764228.1 | - | 2 - 3 | 449 | 64 | |
GQ472229.1 | - | 2 - 3 | 422 | 64 |