Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORBS1
Entrez Gene ID
10580
Gene Name
sorbin and SH3 domain containing 1
Gene Aliases
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location
Chr.10: 95311773 - 95561420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034954.1 NP_001030126.1 9 - 10 1028 75
NM_001034955.1 NP_001030127.1 9 - 10 1028 75
NM_001034956.1 NP_001030128.1 9 - 10 821 75
NM_001034957.1 NP_001030129.1 6 - 7 536 75
NM_001290294.1 NP_001277223.1 11 - 12 1215 75
NM_001290295.1 NP_001277224.1 9 - 10 750 75
NM_001290296.1 NP_001277225.1 8 - 9 1001 75
NM_001290297.1 NP_001277226.1 7 - 8 563 75
NM_001290298.1 NP_001277227.1 7 - 8 632 75
NM_006434.2 NP_006425.2 6 - 7 536 75
NM_015385.3 NP_056200.1 10 - 11 1119 75
NM_024991.2 NP_079267.1 10 - 11 846 75
XM_006717589.1 XP_006717652.1 10 - 11 1141 75
XM_006717593.1 XP_006717656.1 8 - 9 701 75
XM_011539140.1 XP_011537442.1 12 - 13 1885 75
XM_011539150.1 XP_011537452.1 12 - 13 1885 75
XM_011539155.1 XP_011537457.1 10 - 11 1787 75
XM_011539167.1 XP_011537469.1 9 - 10 1070 75
XM_017015500.1 XP_016870989.1 12 - 13 1885 75
XM_017015501.1 XP_016870990.1 12 - 13 1885 75
XM_017015502.1 XP_016870991.1 12 - 13 1885 75
XM_017015503.1 XP_016870992.1 12 - 13 1885 75
XM_017015504.1 XP_016870993.1 10 - 11 799 75
XM_017015505.1 XP_016870994.1 12 - 13 1885 75
XM_017015506.1 XP_016870995.1 10 - 11 799 75
XM_017015507.1 XP_016870996.1 12 - 13 1885 75
XM_017015508.1 XP_016870997.1 10 - 11 799 75
XM_017015509.1 XP_016870998.1 9 - 10 728 75
XM_017015510.1 XP_016870999.1 12 - 13 1885 75
XM_017015511.1 XP_016871000.1 10 - 11 799 75
XM_017015512.1 XP_016871001.1 8 - 9 701 75
XM_017015513.1 XP_016871002.1 8 - 9 701 75
XM_017015514.1 XP_016871003.1 8 - 9 701 75
XM_017015515.1 XP_016871004.1 11 - 12 1168 75
XM_017015516.1 XP_016871005.1 10 - 11 1141 75
XM_017015517.1 XP_016871006.1 11 - 12 1168 75
XM_017015518.1 XP_016871007.1 10 - 11 1097 75
XM_017015519.1 XP_016871008.1 9 - 10 1070 75
XM_017015520.1 XP_016871009.1 11 - 12 1168 75
XM_017015521.1 XP_016871010.1 10 - 11 1097 75
XM_017015522.1 XP_016871011.1 10 - 11 1141 75
XM_017015523.1 XP_016871012.1 10 - 11 1097 75
XM_017015524.1 XP_016871013.1 9 - 10 1070 75
XM_017015525.1 XP_016871014.1 9 - 10 728 75
XM_017015526.1 XP_016871015.1 10 - 11 799 75
XM_017015527.1 XP_016871016.1 9 - 10 728 75
XM_017015528.1 XP_016871017.1 9 - 10 772 75
XM_017015529.1 XP_016871018.1 10 - 11 799 75
XM_017015530.1 XP_016871019.1 10 - 11 799 75
XM_017015531.1 XP_016871020.1 9 - 10 728 75
XM_017015532.1 XP_016871021.1 9 - 10 772 75
XM_017015533.1 XP_016871022.1 8 - 9 701 75
XM_017015534.1 XP_016871023.1 10 - 11 799 75
XM_017015535.1 XP_016871024.1 9 - 10 728 75
XM_017015536.1 XP_016871025.1 9 - 10 728 75
XM_017015537.1 XP_016871026.1 9 - 10 772 75
XM_017015538.1 XP_016871027.1 9 - 10 772 75
XM_017015539.1 XP_016871028.1 8 - 9 701 75
XM_017015540.1 XP_016871029.1 8 - 9 701 75
GenBank mRNA AB037717.1 - 10 - 11 778 75
AF136380.1 - 5 - 6 491 75
AF136381.1 - 7 - 8 956 75
AF330623.1 - 6 - 7 518 75
AF356525.1 - 9 - 10 1039 75
AF356526.1 - 9 - 10 1039 75
AF356527.1 - 9 - 10 832 75
AJ489942.1 - 6 - 7 587 75
AK300407.1 - 9 - 10 703 75
AK304753.1 - 10 - 11 1762 75
AL117472.1 - 10 - 11 1077 75
AM260536.1 - 11 - 12 1173 75
BC042612.1 - 8 - 9 1143 75
BC152463.1 - 10 - 11 778 75

View Full Product Details