Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORBS1
Entrez Gene ID
10580
Gene Name
sorbin and SH3 domain containing 1
Gene Aliases
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location
Chr.10: 95311773 - 95561420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034954.1 NP_001030126.1 4 - 5 343 66
NM_001034955.1 NP_001030127.1 4 - 5 343 66
NM_001034956.1 NP_001030128.1 4 - 5 343 66
NM_001290294.1 NP_001277223.1 6 - 7 530 66
NM_001290295.1 NP_001277224.1 5 - 6 434 66
NM_001290297.1 NP_001277226.1 3 - 4 247 66
NM_015385.3 NP_056200.1 5 - 6 434 66
NM_024991.2 NP_079267.1 6 - 7 530 66
XM_011539140.1 XP_011537442.1 6 - 7 483 66
XM_011539150.1 XP_011537452.1 6 - 7 483 66
XM_017015500.1 XP_016870989.1 6 - 7 483 66
XM_017015501.1 XP_016870990.1 6 - 7 483 66
XM_017015502.1 XP_016870991.1 6 - 7 483 66
XM_017015503.1 XP_016870992.1 6 - 7 483 66
XM_017015504.1 XP_016870993.1 6 - 7 483 66
XM_017015505.1 XP_016870994.1 6 - 7 483 66
XM_017015506.1 XP_016870995.1 6 - 7 483 66
XM_017015507.1 XP_016870996.1 6 - 7 483 66
XM_017015508.1 XP_016870997.1 6 - 7 483 66
XM_017015509.1 XP_016870998.1 5 - 6 412 66
XM_017015510.1 XP_016870999.1 6 - 7 483 66
XM_017015511.1 XP_016871000.1 6 - 7 483 66
XM_017015515.1 XP_016871004.1 6 - 7 483 66
XM_017015517.1 XP_016871006.1 6 - 7 483 66
XM_017015518.1 XP_016871007.1 5 - 6 412 66
XM_017015520.1 XP_016871009.1 6 - 7 483 66
XM_017015521.1 XP_016871010.1 5 - 6 412 66
XM_017015523.1 XP_016871012.1 5 - 6 412 66
XM_017015525.1 XP_016871014.1 5 - 6 412 66
XM_017015526.1 XP_016871015.1 6 - 7 483 66
XM_017015527.1 XP_016871016.1 5 - 6 412 66
XM_017015529.1 XP_016871018.1 6 - 7 483 66
XM_017015530.1 XP_016871019.1 6 - 7 483 66
XM_017015531.1 XP_016871020.1 5 - 6 412 66
XM_017015534.1 XP_016871023.1 6 - 7 483 66
XM_017015535.1 XP_016871024.1 5 - 6 412 66
XM_017015536.1 XP_016871025.1 5 - 6 412 66
GenBank mRNA AB037717.1 - 6 - 7 462 66
AF330623.1 - 2 - 3 202 66
AF356525.1 - 4 - 5 354 66
AF356526.1 - 4 - 5 354 66
AF356527.1 - 4 - 5 354 66
AK022468.1 - 6 - 7 509 66
AK300407.1 - 5 - 6 387 66
AL117472.1 - 5 - 6 392 66
AM260536.1 - 6 - 7 488 66
BC152463.1 - 6 - 7 462 66

View Full Product Details