The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003854.2 | NP_003845.2 | 5 - 6 | 780 | 121 |
XM_005264035.1 | XP_005264092.1 | 5 - 6 | 773 | 121 | |
XM_005264036.1 | XP_005264093.1 | 5 - 6 | 695 | 121 | |
XM_006712820.2 | XP_006712883.1 | 3 - 4 | 484 | 121 | |
XM_006712822.2 | XP_006712885.1 | 2 - 3 | 254 | 121 | |
XM_011512092.1 | XP_011510394.1 | 5 - 6 | 773 | 121 | |
XM_011512094.1 | XP_011510396.1 | 5 - 6 | 695 | 121 | |
XM_011512096.1 | XP_011510398.1 | 5 - 6 | 773 | 121 | |
XM_017005174.1 | XP_016860663.1 | 5 - 6 | 773 | 121 | |
XM_017005178.1 | XP_016860667.1 | 4 - 5 | 526 | 121 | |
XM_017005179.1 | XP_016860668.1 | 4 - 5 | 779 | 121 | |
GenBank mRNA | AF284434.1 | - | 5 - 6 | 788 | 121 |
AK315334.1 | - | 5 - 6 | 695 | 121 | |
U49065.1 | - | 5 - 6 | 788 | 121 |