The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003854.2 | NP_003845.2 | 7 - 8 | 984 | 125 |
XM_005264035.1 | XP_005264092.1 | 7 - 8 | 977 | 125 | |
XM_005264036.1 | XP_005264093.1 | 7 - 8 | 899 | 125 | |
XM_006712820.2 | XP_006712883.1 | 5 - 6 | 688 | 125 | |
XM_006712822.2 | XP_006712885.1 | 4 - 5 | 458 | 125 | |
XM_011512091.1 | XP_011510393.1 | 7 - 8 | 974 | 125 | |
XM_011512092.1 | XP_011510394.1 | 7 - 8 | 977 | 125 | |
XM_011512093.1 | XP_011510395.1 | 6 - 7 | 902 | 125 | |
XM_011512094.1 | XP_011510396.1 | 7 - 8 | 899 | 125 | |
XM_011512096.1 | XP_011510398.1 | 7 - 8 | 977 | 125 | |
XM_017005174.1 | XP_016860663.1 | 7 - 8 | 977 | 125 | |
XM_017005175.1 | XP_016860664.1 | 5 - 6 | 685 | 125 | |
XM_017005176.1 | XP_016860665.1 | 4 - 5 | 613 | 125 | |
XM_017005177.1 | XP_016860666.1 | 6 - 7 | 861 | 125 | |
XM_017005178.1 | XP_016860667.1 | 6 - 7 | 730 | 125 | |
XM_017005179.1 | XP_016860668.1 | 6 - 7 | 983 | 125 | |
XM_017005180.1 | XP_016860669.1 | 4 - 5 | 591 | 125 | |
GenBank mRNA | AF284434.1 | - | 7 - 8 | 992 | 125 |
AK315334.1 | - | 7 - 8 | 899 | 125 | |
BC107064.2 | - | 5 - 6 | 513 | 125 | |
U49065.1 | - | 7 - 8 | 992 | 125 |