The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003854.2 | NP_003845.2 | 9 - 10 | 1263 | 90 |
XM_005264035.1 | XP_005264092.1 | 9 - 10 | 1256 | 90 | |
XM_005264036.1 | XP_005264093.1 | 9 - 10 | 1178 | 90 | |
XM_006712820.2 | XP_006712883.1 | 7 - 8 | 967 | 90 | |
XM_006712822.2 | XP_006712885.1 | 6 - 7 | 737 | 90 | |
XM_011512091.1 | XP_011510393.1 | 9 - 10 | 1253 | 90 | |
XM_011512092.1 | XP_011510394.1 | 9 - 10 | 1256 | 90 | |
XM_011512093.1 | XP_011510395.1 | 8 - 9 | 1181 | 90 | |
XM_011512094.1 | XP_011510396.1 | 9 - 10 | 1178 | 90 | |
XM_017005174.1 | XP_016860663.1 | 9 - 10 | 1256 | 90 | |
XM_017005175.1 | XP_016860664.1 | 7 - 8 | 964 | 90 | |
XM_017005176.1 | XP_016860665.1 | 6 - 7 | 892 | 90 | |
XM_017005177.1 | XP_016860666.1 | 8 - 9 | 1140 | 90 | |
XM_017005178.1 | XP_016860667.1 | 8 - 9 | 1009 | 90 | |
XM_017005179.1 | XP_016860668.1 | 8 - 9 | 1262 | 90 | |
XM_017005180.1 | XP_016860669.1 | 6 - 7 | 870 | 90 | |
GenBank mRNA | AF284434.1 | - | 9 - 10 | 1271 | 90 |
AK315334.1 | - | 9 - 10 | 1178 | 90 | |
BC107064.2 | - | 7 - 8 | 792 | 90 | |
U49065.1 | - | 9 - 10 | 1271 | 90 |