The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022739.3 | NP_073576.1 | 15 - 16 | 2134 | 96 |
XM_005257585.3 | XP_005257642.1 | 14 - 15 | 1734 | 96 | |
XM_011525128.1 | XP_011523430.1 | 15 - 16 | 1752 | 96 | |
GenBank mRNA | AF301463.1 | - | 15 - 16 | 1754 | 96 |
AF310676.1 | - | 15 - 16 | 1746 | 96 | |
AY014180.1 | - | 15 - 16 | 1934 | 96 | |
BC009527.1 | - | 3 - 4 | 368 | 96 | |
BC093876.1 | - | 15 - 16 | 1816 | 96 | |
BC111945.1 | - | 15 - 16 | 1818 | 96 | |
CR936721.1 | - | 16 - 17 | 1869 | 96 |