The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022739.3 | NP_073576.1 | 6 - 7 | 869 | 68 |
XM_005257585.3 | XP_005257642.1 | 5 - 6 | 469 | 68 | |
XM_011525128.1 | XP_011523430.1 | 6 - 7 | 487 | 68 | |
GenBank mRNA | AF301463.1 | - | 6 - 7 | 489 | 68 |
AF310676.1 | - | 6 - 7 | 481 | 68 | |
AY014180.1 | - | 6 - 7 | 669 | 68 | |
BC093876.1 | - | 6 - 7 | 551 | 68 | |
BC111945.1 | - | 6 - 7 | 553 | 68 | |
CR936721.1 | - | 7 - 8 | 604 | 68 |